Learn More
Lenz Laborglasintrumente Cones
Product Code. 10254001
Click to view available options
Joint Size:
NS 40/38
NS 5/13
NS 50/42
NS 55/44
NS 60/46
NS 7/16
NS 71/51
NS 85/55
Unit Size:
Each
Pack of 10
Product Code. 10254001
Brand: Lenz Laborglasintrumente 01100240
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12249 form B and in addition to standard
  • Conical ground cones
  • Made of DURAN™ tubing
Specifications
Quantity 10/Pk.
Joint Size NS 40/38
Thickness (Metric) Wall 2 mm
Diameter (Metric) Outer Tube 36 mm
Material Borosilicate Glass (3.3)