Learn More
BOLA Conductors for Magnetic Stirrer Heads
Click to view available options
Type:
Support Rod
Unit Size:
Each
Description
- Borosilicate glass with ground joint and glued upper bearing.
- Alternatively without ground joint for fusing on the reactor lid
- Upper bearing to be glued is included with Ground Joint NS 29/32
Specifications
Specifications
| Type | Support Rod |
| Material | Glass |