Learn More
Hanna Instruments™ Conductivity/Tds/Salinity/Temperature Meter
Brand:  Hanna Instruments™ HI-98360
Product Code. 11687240
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Handheld with HI76309/1.5 probe with 1.5m cable, batteries, carry case and instructions TDS 0 to 1499ppm(mg/L),1.5 to 400g/L,NaCl 400% temp 9.9 - 120°C, EC 0 to 2999μS/cm, 3 to 500mS/cm Hanna