missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standards NIST Traceable, 100000μS/cm, Reagecon™
Click to view available options
Quantity:
500 mL
Description
- Accurate to ±1%
- Traceable to NIST
- GLP compliant
- Certificate of analysis supplied with each bottle (including method of analysis and standard reference material detail)
- Wide range: 20 values from 1.3μS/cm to 500,000μS/cm inclusive
- An enhanced shelf life is assured through the use of proprietary technologyAccurate conductivity measurement requires periodic calibration of the conductivity sensor in order to determine its cell constant (the multiplier used to convert the sensor's output to the displayed conductivity). Our standard value conductivity standards match the pre-programmed values used by all leading brands of conductivity meters for automatic calibration, While the broad range standards allow flexibility.
Specifications
Quantity | 500 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'