missing translation for 'onlineSavingsMsg'
Learn More

Basic Line Conductivity Standard, 1.3μS/cm, Hamilton™

Product Code. 30067371
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30067371 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30067371 Supplier Hamilton Supplier No. 238973
  • ONLINE EXCLUSIVE PRICE
    £160.22
    £147.53 / 250mL
    Save £12.69
    7% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 1.3μS/cm comes with a certificate with calibration document form DFM.

  • Remains stable for up to 1 year
  • Expiration date shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes

Specifications

Packaging Glass bottle
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.