missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard Solution, 25μS/cm, Hach Lange™
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Specifications
Specifications
| Methods | Standard solution verified by Reference Materials Laboratory |
| Analyte or Component Names | Sodium Chloride |
| Conductivity | 25.0 μS/cm ±5% at 25°C |
| Physical Form | Liquid |
| Concentration | 25.0 μS/cm ±5% at 25°C |
| Element | Sodium Chloride |
| Quantity | 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction