missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, KCl, SPEX CertiPrep™

Product Code. 15266386 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15266386

Brand: SPEX Certiprep TDS2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Conductivity1000μS/cm

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7447-40-7, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.07, 99.93
Analyte or Component Names KCL
Molecular Formula ClK
MDL Number MFCD00011360
Technique Calibration of Conductivity Instrument
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Type Certified Reference Material
SMILES [Cl-].[K+]
Element KCL
IUPAC Name potassium;chloride
Solvent or Matrix Water
Molecular Weight (g/mol) 74.55
PubChem CID 4873
Quantity 500 mL
ChEBI CHEBI:32588
Concentration or Composition (by Analyte or Components) 650mg/L
Chemical Name or Material Conductivity Standard, Total Dissolved Solids as Potassium Chloride
Show More Show Less

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.