missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard at 25°C, NIST Traceable, 500 μS/cm, Reagecon™

Product Code. 17953792 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
5 L
Unit Size:
250mL
5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953792 500 mL 500mL
16522350 250 mL 250mL
16512350 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953792 Supplier Reagecon Supplier No. CRMCSKC500
  • ONLINE EXCLUSIVE PRICE
    £262.00
    £256.54 / 500mL
    Save £5.46
    2% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Tested and certified by an ISO17025 Accredited Test Method (A2LA Ref: 6739.03) and NIST traceable.

  • Reagecon offers high quality Conductivity Standards that support accurate conductivity measurements and can be used for sensor calibration, quality control, method validation, and instrument qualification.
  • The standards are aqueous-based, minimising errors caused by matrix mismatch.
  • Directly traceable to NIST reference materials, meeting the specification requirements of all major Pharmacopoeias.
  • Provided with a comprehensive Certificate of Analysis.
  • A temperature conductivity table can be accessed via the product label and clearly displayed lot numbers and expiry dates for ease of use.
  • A low temperature coefficient reduces temperature related errors, allowing reliable measurements without strict temperature control and supporting field use.
  • High accuracy allows the standards function effectively as both calibrators and controls for demanding industrial applications.
  • Manufactured to stringent specifications, ensuring extended shelf life and stability.

Specifications

Chemical Name or Material Conductivity Standard
Packaging HDPE Bottle
Conductivity 500 μS/cm ±1.0%
Type Certified Reference Material
Molecular Weight (g/mol) Unknown
Quantity 500 mL
Grade Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.