Learn More
Basic Line Conductivity Standard, 5μS/cm, Hamilton™
Product Code. 10747413
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
Each
Product Code. Quantity unitSize
10747413 250 mL Each
1 options
Product Code. 10747413 Supplier Hamilton Supplier No. 238926
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 5μS/cm comes with a certificate with calibration document form DFM.
  • Remains stable for up to 3 years
  • Expiration date is shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes
Specifications
Packaging Glass bottle
Quantity 250 mL