missing translation for 'onlineSavingsMsg'
Learn More

Conductivity solution 718μS/cm, Fisher chemical™
BIOPHARMA

Product Code. 12914495 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12914495 500 mL Bottle 500mL
12904495 1000 mL Bottle 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12914495 Supplier Fisher Chemical Supplier No. CTE 007 500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Conductivity 718μS/cm
Type Conductivity Solution
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.