missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Solution, 1413μS.cm-1, Fisher Chemical™

Product Code. 12944495 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
Product Code. Quantity unitSize
12944495 1000 mL 1000mL
12954495 500 mL 500mL
2 options
This item is not returnable. View return policy

Product Code. 12944495

Brand: Fisher Chemical CTE 009 1L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Conductivity 1413μS.cm-1
Physical Form Liquid
Quantity 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.