missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Conductivity Flow Cell

Brand:  Cytiva 18-1114-43

Code : GX

Additional Details : Weight : 0.21500kg

 View more versions of this product

Product Code. 11320242

  • £708.00 / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

M6
Titanium, Ethylene propylene diene and Fluoro plastics
Conductivity Flow Cell
1 Pack
Conductivity Monitor
50 bar
0.014 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers