Learn More
Cytiva Conductivity Cell
Product Code. 15289774 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
ÄKTA Start
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15289774 ÄKTA Start Each
1 options
Product Code. 15289774
Brand: Cytiva 29024021
Only null left
Add to basket
Add to basket
For use in protein purification applications
For use with the AKTA start automated protein purification system. The conductivity monitor continuously reads the conductivity of the liquid in the conductivity flow cell. The conductivity value is automatically calculated by multiplying the measured conductance by the cell constant of the flow cell, and taking the temperature effect into account. The cell constant is factory calibrated and a built-in temperature sensor continuously measures the temperature of the liquid in the flow cell.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Protein Purification
For Use With (Equipment) ÄKTA Start
Type Conductivity Cell
Quantity 1 Each