Learn More
Conductivity Calibration Standards, YSI™
Description
- Conductivity solutions are inspected with reference to primary standard solutions according to OIML recommendation 56
- Bottles include a table of corrections at temperatures between 20 and 30°C
- Available as: 1 Quart or in a box of 8 individual pints
Specifications
Specifications
| Packaging | Glass Bottle |
| Conductivity | 100,000μS/cm |
| Type | Conductivity Calibration Standard |
| Quantity | 1 qt. |