missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ 50μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Description
- Fine, thin tip facilitates dosing of micro volumes
- Usable with 1000μL pipetting channels/CO-RE 96 Probe Head
- Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Specifications
Specifications
| Volume (Metric) | 50 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Black |
| For Use With (Application) | Enhanced pipetting safety & reliability with the CO-RE PerfectTouch system |
| Tip Style | Conductive |
| Disposable | Single-use |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?