missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Conditioning Reagent Dx

Product Code. 15869846 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869846 1 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869846 Supplier Applied Biosystems™ Supplier No. 4409543
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Conditioning Reagent Dx is designed for use with Applied Biosystems Dx genetic analyzers.

This Conditioning Reagent Dx is designed for use withApplied Biosystems Dx genetic analyzers.

Conditioning Reagent Dx is available as a ready-to-use pouch and is used for priming the polymer pump, washing the polymer pump between polymer type changes, and during instrument shut down. The pouch has adequate volume for a one-time use. The top part of the pouch fitment is sealed with a plastic film, which should be removed prior to installation on to the instrument.

This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.

The 3500 Dx series conditioning reagent, anode buffer, cathode buffer and septa cathode buffer are required for instrument operation.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Starting Material DNA, RNA
Content And Storage Store at 2 - 8°C.
Format Pouch
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
Product Type Conditioning Reagent
Quantity 1 Pk.
Shipping Condition Wet Ice

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.