Learn More
BUCHI Condensate Trap for Rotavapor Condenser
Product Code. 15570570 Shop All Buchi Products
Change view
Click to view available options
Material:
Thermoplastic
Unit Size:
Each
Product Code. Material Unit Size
15570570 Thermoplastic Each
1 options
Product Code. 15570570 Supplier BUCHI Supplier No. 11061985
  • ONLINE EXCLUSIVE PRICE
    £11.95
    £10.00 / Pack of 1
    Save £1.95
    16% Off
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Collects and drains condensate, which may accumulate on the outside of the condenser.
  • For use with V (vertical coil) and HP (high-performance) condensers
  • Green
  • TPE
  • Drain hose connector diameter: 8 mm
TRUSTED_SUSTAINABILITY
Specifications
Material Thermoplastic
Type Condenser Insert