missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Crystal Violet Stain, Concentrated
Click to view available options
Quantity:
100mL
Unit Size:
100mL
Description
Pro-Lab Diagnostics™ stains are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all stains offer excellent performance characteristics.
- 10X concentrated – 100mL is diluted to 1L of ready-to-use stain
- Crystal Violet is the primary stain of the Gram staining method
- Stains the cell walls of organisms and is retained in Gram-positive bacteria after decolorization
Specifications
Specifications
| Solution Type | Concentrated Reagent |
| Type | Crystal Violet |
| Quantity | 100mL |
| Certifications/Compliance | (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark |
| Format | Liquid |
| For Use With (Application) | For Gram staining |
| Color | Dark violet |
| Composition | Ethanol, C.I. basic violet 3, Methanol |
| Storage Requirements | <20°C |
| Stain Type | Gram Stain |
| Show More |
Safety and Handling
Product Identifier
- Crystal Violet Concentrate
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H226-Flammable liquid and vapour.
- H332-Harmful if inhaled.
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H371-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
- P201-Obtain special instructions before use.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P370+P378-In case of fire: Use to extinguish.
- P391-Collect spillage.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- MIXTURE LIST-Contains : ethanol, C.I. basicviolet 3, methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction