missing translation for 'onlineSavingsMsg'
Learn More

Concanavalin A

Product Code. 16821900
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16821900 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16821900 Supplier Thermo Scientific Alfa Aesar Supplier No. J61221.MC
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

A plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts | CAS: 11028-71-0

Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Concanavalin A is a plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.

Solubility
Slightly soluble in water and phosfate buffered saline.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11028-71-0
MDL Number MFCD00071069
Synonym ConA

Specifications

Type Concanavalin A
Physical Form Lyophilized Powder
Color White
Quantity 100 mg
Merck Index 14,2495
Solubility Information Slightly soluble in water and phosfate buffered saline.
Chemical Name or Material Concanavalin A

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.