missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COMT Monoclonal Antibody (C4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551130
This item is not returnable.
View return policy
Description
Catechol-O-methyltransferase catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines, including the neurotransmitters dopamine, epinephrine, and norepinephrine. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to its role in the metabolism of endogenous substances, COMT is important in the metabolism of catechol drugs used in the treatment of hypertension, asthma, and Parkinson disease. COMT is found in two forms in tissues, a soluble form (S-COMT) and a membrane-bound form (MB-COMT). The differences between S-COMT and MB-COMT reside within the N-termini. Several transcript variants are formed through the use of alternative translation initiation sites and promoters.
Specifications
| COMT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P22734 | |
| Comt | |
| Recombinant Rat COMT protein, Gly63-Ser264 (Accession #P22734). | |
| 20 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C4 | |
| Unconjugated | |
| Comt | |
| Catechol O-methyltransferase; catecholamine-O-methyltransferase; catechol-O-methyltransferase; catechol-O-methyltransferase 1; catechol-O-methyltransferase isoform; Comt; Comt1; D16Wsu103e; D330014B15Rik; epididymis secretory sperm binding protein Li 98n; HEL-S-98n; OTTHUMP00000197749; OTTHUMP00000197750; testicular tissue protein Li 42 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 24267 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction