missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ COMT Antibody Blocking Peptide

Product Code. 18559681 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
This item is not returnable. View return policy

Product Code. 18559681

Brand: Novus Biologicals™ NB1001780PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene ID (Entrez) 1312
Species Human
Purity >50%, by HPLC
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-1780
Gene Alias catechol O-methyltransferase, catechol-O-methyltransferase, EC 2.1.1.6
Gene Symbol COMT
Quantity 0.1 mg
Research Category Lipid and Metabolism
Storage Buffer Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.