missing translation for 'onlineSavingsMsg'
Learn More

Comprehensive Mix B Standard: Au, B, Be, Co, Cr, Cu, Fe, Ge, Hf, Ir, K, Li, Mn, Mo, Nb, Ni, Os, Pd, Pt, Re, Rh, Ru, Sb, Si, Sn, Ta, Te, Ti, V, W, Zn, Zr @ 10 μg/mL in 40% Aqua Regia, tr. HF, VHG™

Product Code. p-200131579
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30003089

Brand: VHG Labs VHGSM90C250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Comprehensive Mix B Standard: Au, B, Be, Co, Cr, Cu, Fe, Ge, Hf, Ir, K, Li, Mn, Mo, Nb, Ni, Os, Pd, Pt, Re, Rh, Ru, Sb, Si, Sn, Ta, Te, Ti, V, W, Zn, Zr @ 10 μg/mL in 40% Aqua Regia, tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antimony/Beryllium/Boron/Chromium/Cobalt/Copper/Germanium/Gold/Hafnium/Iridium/Iron/Lithium/Manganese/Molybdenum/Nickel/Niobium/Osmium/Palladium/Platinum/Potassium/Rhenium/Rhodium/Ruthenium/Silicon/Tantalum/Tellurium/Tin/Titanium/Tungsten/Vanadium/Zinc/Zirconium
CAS 7440-36-0, 7440-41-7, 7440-42-8, 7440-47-3, 7440-48-4, 7440-50-8
Methods ICP/ICP-MS
Analyte or Component Names Antimony/Beryllium/Boron/Chromium/Cobalt/Copper/Germanium/Gold/Hafnium/Iridium/Iron/Lithium/Manganese/Molybdenum/Nickel/Niobium/Osmium/Palladium/Platinum/Potassium/Rhenium/Rhodium/Ruthenium/Silicon/Tantalum/Tellurium/Tin/Titanium/Tungsten/Vanadium/Zinc/Zirconium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute Aqua Regia
Quantity 250 mL
Concentration or Composition (by Analyte or Components) Antimony 10 μg/mL/Beryllium 10 μg/mL/Boron 10 μg/mL/Chromium 10 μg/mL/Cobalt 10 μg/mL/Copper 10 μg/mL/Germanium 10 μg/mL/Gold 10 μg/mL/Hafnium 10 μg/mL/Iridium 10 μg/mL/Iron 10 μg/mL/Lithium 10 μg/mL/Manganese 10 μg/mL/Molybdenum 10 μg/mL/Nickel 10 μg/mL/Niobium 10 μg/mL/Osmium 10 μg/mL/Palladium 10 μg/mL/Platinum 10 μg/mL/Potassium 10 μg/mL/Rhenium 10 μg/mL/Rhodium 10 μg/mL/Ruthenium 10 μg/mL/Silicon 10 μg/mL/Tantalum 10 μg/mL/Tellurium 10 μg/mL/Tin 10 μg/mL/Titanium 10 μg/mL/Tungsten 10 μg/mL/Vanadium 10 μg/mL/Zinc 10 μg/mL/Zirconium 10 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.