missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Comprehensive Mix B Standard: Au, B, Be, Co, Cr, Cu, Fe, Ge, Hf, Ir, K, Li, Mn, Mo, Nb, Ni, Os, Pd, Pt, Re, Rh, Ru, Sb, Si, Sn, Ta, Te, Ti, V, W, Zn, Zr @ 10 μg/mL in 40% Aqua Regia, tr. HF, VHG™
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Description
Comprehensive Mix B Standard: Au, B, Be, Co, Cr, Cu, Fe, Ge, Hf, Ir, K, Li, Mn, Mo, Nb, Ni, Os, Pd, Pt, Re, Rh, Ru, Sb, Si, Sn, Ta, Te, Ti, V, W, Zn, Zr @ 10 μg/mL in 40% Aqua Regia, tr. HF
Specifications
Specifications
| Chemical Name or Material | Antimony/Beryllium/Boron/Chromium/Cobalt/Copper/Germanium/Gold/Hafnium/Iridium/Iron/Lithium/Manganese/Molybdenum/Nickel/Niobium/Osmium/Palladium/Platinum/Potassium/Rhenium/Rhodium/Ruthenium/Silicon/Tantalum/Tellurium/Tin/Titanium/Tungsten/Vanadium/Zinc/Zirconium |
| CAS | 7440-36-0, 7440-41-7, 7440-42-8, 7440-47-3, 7440-48-4, 7440-50-8 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Antimony/Beryllium/Boron/Chromium/Cobalt/Copper/Germanium/Gold/Hafnium/Iridium/Iron/Lithium/Manganese/Molybdenum/Nickel/Niobium/Osmium/Palladium/Platinum/Potassium/Rhenium/Rhodium/Ruthenium/Silicon/Tantalum/Tellurium/Tin/Titanium/Tungsten/Vanadium/Zinc/Zirconium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute Aqua Regia |
| Quantity | 250 mL |
| Concentration or Composition (by Analyte or Components) | Antimony 10 μg/mL/Beryllium 10 μg/mL/Boron 10 μg/mL/Chromium 10 μg/mL/Cobalt 10 μg/mL/Copper 10 μg/mL/Germanium 10 μg/mL/Gold 10 μg/mL/Hafnium 10 μg/mL/Iridium 10 μg/mL/Iron 10 μg/mL/Lithium 10 μg/mL/Manganese 10 μg/mL/Molybdenum 10 μg/mL/Nickel 10 μg/mL/Niobium 10 μg/mL/Osmium 10 μg/mL/Palladium 10 μg/mL/Platinum 10 μg/mL/Potassium 10 μg/mL/Rhenium 10 μg/mL/Rhodium 10 μg/mL/Ruthenium 10 μg/mL/Silicon 10 μg/mL/Tantalum 10 μg/mL/Tellurium 10 μg/mL/Tin 10 μg/mL/Titanium 10 μg/mL/Tungsten 10 μg/mL/Vanadium 10 μg/mL/Zinc 10 μg/mL/Zirconium 10 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction