missing translation for 'onlineSavingsMsg'
Learn More

Compound E, MedChemExpress

Product Code. 16473406
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473406 25 mg 25mg
16468597 5 mg 5mg
16465978 10 mg 10mg
16451399 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473406 Supplier Medchem Express Supplier No. HY14176
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Compound E is a γ-secretase inhibitor. Compound E bloks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage with IC50s of 0.24, 0.37, 0.32 nM, respectively.

Compound E is a γ-secretase inhibitor. Compound E bloks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage with IC50s of 0.24, 0.37, 0.32 nM, respectively.

Specifications

Chemical Name or Material Compound E
CAS 209986-17-4
Color White
Purity Grade Notes Research
Molecular Formula C27H24F2N4O3
Quantity 25 mg
Synonym γ-Secretase-IN-1
Solubility Information DMSO : 115 mg/mL (234.45 mM; ultrasonic and warming and heat to 60°C)
SMILES FC1=CC(CC(N[C@@H](C)C(N[C@@H]2C(N(C)C(C=CC=C3)=C3C(C4=CC=CC=C4)=N2)=O)=O)=O)=CC(F)=C1
Molecular Weight (g/mol) 490.5
Formula Weight 490.5
Percent Purity 98.75%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.