missing translation for 'onlineSavingsMsg'
Learn More

Dolasetron, MedChemExpress

Product Code. 16468598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16468598

Brand: Medchem Express HYB0750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dolasetron(MDL-73147) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy.

Dolasetron(MDL-73147) is a serotonin 5-HT3 receptor antagonist used to treat nausea and vomiting following chemotherapy.

Specifications

Chemical Name or Material Dolasetron
CAS 115956-12-2
Color White
Purity Grade Notes Research
Molecular Formula C19H20N2O3
Quantity 10 mg
Synonym MDL-73147
Solubility Information DMSO : ≥ 300 mg/mL (924.87 mM)
SMILES O=C(C1=CNC2=C1C=CC=C2)O[C@@H]3C[C@@](CC4C5)([H])[N@](CC4=O)[C@@]5([H])C3
Molecular Weight (g/mol) 324.37
Formula Weight 324.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.