missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Compound E

Product Code. 17110302
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17110302 1 mg 1mg
17100302 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17110302 Supplier STEMCELL Technologies Supplier No. 73954
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Notch pathway inhibitor; Inhibits Notch receptor and amyloid precursor protein

  • Compound E is a potent, cell-permeable, selective inhibitor that blocks the cleavage of both gamma-secretase and the Notch intracellular domain with similar IC50 values, ranging from 0.24 to 0.37 nM (Beher et al.; Seiffert et al.)
  • Inhibits growth, differentiation, and motility of neuroblastoma cells (Ferrari-Toninelli et al.)
  • Accelerates the differentiation of human embryonic stem cells into primitive neural stem cells (Li et al.)

Specifications

CAS 209986-17-4
Synonym γ-secretase inhibitor XXI
Quantity 1 mg
Molecular Formula C27H24F2N4O3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.