missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complexin-2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£143.00
Specifications
| Antigen | Complexin-2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Complexin-2 Polyclonal antibody specifically detects Complexin-2 in Human samples. It is validated for ELISASpecifications
| Complexin-2 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Neuroscience, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 10814 | |
| IgG | |
| Antigen and protein A Affinity-purified | |
| Antibody |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| complexin 2, Complexin II, complexin-2, CPX II, CPX2, CPX-2, DKFZp547D155, Hfb1, MGC138492, synaphin 1,921-L, synaphin-1 | |
| Produced in rabbits immunized with purified, recombinant Human CPLX2 (Q6PUV4-1; Asp2-Lys134) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title