missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complexin-2 Antibody (021), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90019CL1
This item is not returnable.
View return policy
Description
Complexin-2 Monoclonal antibody specifically detects Complexin-2 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry/Immunofluorescence
Specifications
| Complexin-2 | |
| Monoclonal | |
| CoraFluor 1 | |
| complexin 2, Complexin II, complexin-2, CPX II, CPX2, CPX-2, DKFZp547D155, Hfb1, MGC138492, synaphin 1,921-L, synaphin-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Complexin-2 (Uniprot#: Q6PUV4-1; Asp2-Lys134). | |
| 0.1 mL | |
| Immunology, Innate Immunity, Neuroscience, Signal Transduction | |
| 10814 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 021 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction