missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complexin-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Tekniske data
| Antigen | Complexin-1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18397196
|
Novus Biologicals
NBP3-03264-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352156
|
Novus Biologicals
NBP3-03264-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Complexin-1 Polyclonal specifically detects Complexin-1 in Human, Mouse, Rat samples. It is validated for Western Blot.Tekniske data
| Complexin-1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| complexin 1, Complexin I, complexin-1, CPX I, CPX1, CPX-I, synaphin 2, Synaphin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of human Complexin-1 (NP_006642.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS with 50% glycerol, pH7.3. | |
| 10815 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel