missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX5A Monoclonal Antibody (6E9B12D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 459120
This item is not returnable.
View return policy
Description
Positive control: Isolated mitochondria from Human heart, Bovine heart, Rat heart, Mouse heart, and HepG2, Human embryonic lung-derived fibroblasts (strain MRC5), HeLa cells.
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The mitochondrially-encoded subunits perform the electron transfer of proton pumping activities. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. This gene encodes the nuclear-encoded subunit Va of the human mitochondrial respiratory chain enzyme. A pseudogene COX5AP1 has been found in chromosome 14q22.
Specifications
| COX5A | |
| Monoclonal | |
| 1 mg/mL | |
| HEPES buffered saline with 0.02% sodium azide | |
| P00426, P11240, P12787, P20674 | |
| COX5A | |
| Bovine COX5A. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 6E9B12D5 | |
| Unconjugated | |
| COX5A | |
| AA959768; CcOX; COX; Cox5a; COX5A protein; COX-VA; cytochrome c oxidase polypeptide Va; cytochrome c oxidase polypeptide, mitochondrial; cytochrome c oxidase subunit 5A; cytochrome c oxidase subunit 5A, mitochondrial; cytochrome c oxidase subunit Va; cytochrome c oxidase, subunit Va; mitochondrial cytochrome c oxidase subunit 5a; mitochondrial cytochrome c oxidase subunit Va; VA | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 12858, 252934, 444878, 9377 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction