missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Complex III Monoclonal Antibody (11A51H12)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ 439400

Product Code. 10338433

  • £479.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Positive control: Bovine heart and Human mitochondria.

Complex III, or ubiquinol cytochrome c oxidoreductase, is a complex of 11 subunits and catalyses electron transfer from reduced ubiquinone or coenzyme Q 10 to cytochrome c. The complex contains 11 different subunits, one of which, cytochrome b, is encoded on mitochondrial DNA, while the remainder subunits are nuclear-encoded. Genetic complex III deficiencies occur but are rare.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Complex III
Monoclonal
1 mg/mL
HEPES buffered saline with 0.02% sodium azide
O14949, O14957, P00125, P00126, P00129, P00130, P00156, P07552, P07919, P08574, P13271, P13272, P14927, P22695, P23004, P31800, P31930, P47985, Q9UDW1
CYC1, MT-CYB, UQCR10, UQCR11, UQCRB, UQCRC1, UQCRC2, UQCRFS1, UQCRH, UQCRQ
Bovine Heart Complex III.
100 μg
Primary
Human, Bovine
Antibody
IgG1 κ
Immunoprecipitation
11A51H12
Unconjugated
CYC1
0710008D09Rik; 1100001F06Rik; 1110020P15Rik; 1110032G10Rik; 1500004O06Rik; 1500040F11Rik; 2210415M14Rik; 2210416J04Rik; 2310021J10Rik; 2610002H19Rik; 2610041P16Rik; 4430402G14Rik; 5830407L17Rik; 8 kDa subunit 9; 9.5kDa; AA408921; AA959903; AA960494; AI875505; AL022707; c1502; COB; Complex III subunit 1; complex III subunit 10; Complex III subunit 2; Complex III subunit 3; Complex III subunit 4; complex III subunit 5; Complex III subunit 6; complex III subunit 7; Complex III subunit 8; complex III subunit 9; Complex III subunit III; Complex III subunit IV; Complex III subunit IX; Complex III subunit VII; complex III subunit VIII; Complex III subunit X; complex III subunit XI; Core protein I; core protein II; CYC1; CYTB; Cytochrome b; cytochrome bc-1 complex core protein II; Cytochrome b-c1 complex subunit 1, mitochondrial; cytochrome b-c1 complex subunit 10; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 2, mitochondrial; Cytochrome b-c1 complex subunit 3; Cytochrome b-c1 complex subunit 4; cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 6, mitochondrial; cytochrome b-c1 complex subunit 7; Cytochrome b-c1 complex subunit 8; cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; cytochrome c1; cytochrome c-1; cytochrome c1 non-heme 11 kDa protein; Cytochrome c1 non-heme 7 kDa protein; cytochrome c1, heme protein, mitochondrial; cytochrome C1, nonheme 11 kDa protein; cytochrome C1, nonheme 7kDa protein; D3S3191; HSPC051; HSPC119; HSPC151; hypothetical protein LOC613899; hypothetical protein LOC616871; liver regeneration-related protein LRRGT00195; Low molecular mass ubiquinone-binding protein; low molecular mass ubiquinone-binding protein (9.5kD); low molecular mass ubiquinone-binding protein (Qpc-pending); LRRGT00195; MC3DN3; MC3DN4; MC3DN5; MC3DN6; Mitochondrial hinge protein; mitochondrial ubiquinone-binding protein; mitochondrially encoded cytochrome b; MTCYB; MT-CYB; QCR1; QCR10; QCR2; QCR6; QCR7; QCR8; QCR9; QPC; QP-C; Rieske iron-sulfur; Rieske iron-sulfur protein; Rieske iron-sulfur protein precursor; Rieske protein UQCRFS1; RIP1; RIS1; RISP; similar to ubiquinol-cytrochrome-c reductase (subunit II); smallest subunit of ubiquinol-cytochrome c reductase; Su9; ubiquinol cytochrome c reductase core protein 2; ubiquinol cytochrome c reductase hinge protein; ubiquinol cytochrome c reductase, 6.4kDa subunit; ubiquinol--cytochrome c reductase; ubiquinol-cytochrome c reductase (6.4kD) subunit; Ubiquinol-cytochrome c reductase 8 kDa protein; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase binding protein-like; ubiquinol-cytochrome c reductase complex (7.2 kD); ubiquinol-cytochrome c reductase complex 11 kDa protein; Ubiquinol-cytochrome c reductase complex 14 kDa protein; ubiquinol-cytochrome c reductase complex 6.4 kDa protein; Ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase complex 7.2kDa protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase complex III subunit VII; ubiquinol-cytochrome c reductase complex protein; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase core protein 1; ubiquinol-cytochrome c reductase core protein 2; ubiquinol-cytochrome c reductase core protein I; ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome c reductase hinge protein; ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase subunit; ubiquinol-cytochrome c reductase, 6.4kDa subunit; ubiquinol-cytochrome c reductase, complex III subunit VI; ubiquinol-cytochrome c reductase, complex III subunit VII; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; ubiquinol-cytochrome c reductase, complex III subunit VIII
Mouse
Ammonium Sulfate Precipitation
RUO
10975, 1537, 27089, 281570, 282393, 282394, 286885, 287020, 29796, 4519, 512500, 613899, 616109, 616871, 7381, 7384, 7385, 7386, 7388
4°C, do not freeze
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.