missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Complete Tubing Kit
Shop All Cytiva Products
Click to view available options
For Use With:
AKTA Start Protein Purification System
Unit Size:
1 set
Description
Contains the complete set of tubing connections for the AKTA start protein purification system.
Specifications
Specifications
| For Use With | AKTA Start Protein Purification System |
| Type | Complete Tubing Kit |
| Includes | Complete set of tubing connections |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction