Learn More
Cytiva Complete Tube Racks
Product Code. p-8001011 Shop All Cytiva Products
Click to view available options
Positions:
40
95
175
Unit Size:
Each
Product Code. 11380302
Brand: Cytiva 19868403
Only null left
Add to basket
Add to basket
For use with Fraction collector F9-R and Fraction collector 920
Includes bowl, tube support, holder and guide. For use with Fraction collector F9-R and Fraction collector 920.
TRUSTED_SUSTAINABILITY
Specifications
Includes Includes bowl, tube support, holder and guide
Product Type Tube Rack
Holds Tubes (12 mm)
Positions 175
For Use With (Equipment) For use with Fraction collector F9-R and Fraction collector 920
Quantity 1 Each
Diameter (Metric) Opening 12 mm