missing translation for 'onlineSavingsMsg'
Learn More

Complement iC3b Antibody (9), Novus Biologicals™

Product Code. 18322162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322162 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322162 Supplier Novus Biologicals Supplier No. NBP311285

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Complement iC3b Monoclonal specifically detects Complement iC3b in Human samples. It is validated for Immunoprecipitation, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement iC3b
Applications Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone 9
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Immunoprecipitation, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Complement C3dg fragment, Complement C3g fragment, Complement fragment iC3b, iC3b, Inactivated C3b
Host Species Rat
Immunogen The monocolonal antibody 9 (also known as YB2/90-5-20) reacts with a neoantigen on iC3, iC3b, C3dg and C3g.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.