missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor MASP3/MASP1 Antibody (2B11), Novus Biologicals™

Product Code. 18338112 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338112 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338112 Supplier Novus Biologicals Supplier No. NBP311245

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Factor MASP3/MASP1 Monoclonal specifically detects Complement Factor MASP3/MASP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor MASP3/MASP1
Applications Western Blot, Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone 2B11
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Complement factor MASP-3, Complement-activating component of Ra-reactive factor, CRARF1, CRARFMAP1, DKFZp686I01199, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.104, EC 3.4.21.42, FLJ26383, mannan-binding lectin serine peptidase 1 (C4/C2 activating component ofRa-reactive factor), mannan-binding lectin serine protease 1, mannan-binding lectin serine protease 1 (C4/C2 activating component ofRa-reactive factor), Mannose-binding lectin-associated serine protease 1, Mannose-binding protein-associated serine protease, MAp44, MASP, MASP-1, MASP3, MGC126283, MGC126284, PRSS5, Ra-reactive factor serine protease p100, RaRF, Serine protease 5
Host Species Mouse
Immunogen The antibody recognizes the heavy chain common to both MASP-1 and MASP-3.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5648
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.