missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement Factor I Monoclonal Antibody (OX-21)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA170056
This item is not returnable.
View return policy
Description
MA1-70056 detects Complement Factor I in human samples. MA1-70056 has been successfully used in flow cytometry, ELISA, RIA, immunoprecipitation and Western blot procedures. The MA1-70056 immunogen is human complement Factor I.
Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 μg/mL.
Specifications
| Complement Factor I | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P05156 | |
| CFI | |
| Human complement Factor I. | |
| 250 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
| OX-21 | |
| Unconjugated | |
| CFI | |
| AHUS3; ARMD13; C3B/C4B inactivator; C3BINA; C3b-INA; C3b-inactivator; Cfi; complement component factor i; complement component I; complement control protein factor I; complement factor I; Complement factor I heavy chain; Complement factor I light chain; factor i; FI; IF; KAF; Konglutinogen-activating factor; light chain of factor I | |
| Mouse | |
| Protein G | |
| RUO | |
| 3426 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction