missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor I Antibody (OTI7C9), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70997R
This item is not returnable.
View return policy
Description
Complement Factor I Monoclonal specifically detects Complement Factor I in Human samples. It is validated for Western Blot.
Specifications
| Complement Factor I | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CFI | |
| Recombinant protein fragment corresponding to amino acids 340-583 of human Complement Factor I (NP_000195) produced in E.coli. | |
| 0.1 mL | |
| 3426.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI7C9 | |
| Western Blot | |
| AHUS3, C3B/C4B inactivator, C3BINA, C3b-inactivator, complement component I, complement control protein factor I, complement factor I, complement factor I heavy chain, EC 3.4.21, EC 3.4.21.45, FIC3b-INA, I factor (complement), IF, KAF, Konglutinogen-activating factor, light chain of factor I | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction