missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor I Antibody (1B3), Novus Biologicals™

Product Code. 18385798 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385798 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18385798 Supplier Novus Biologicals Supplier No. H00003426M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Factor I Monoclonal antibody specifically detects Complement Factor I in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor I
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1B3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA 1:100 to 1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000195
Gene Alias AHUS3, C3B/C4B inactivator, C3BINA, C3b-inactivator, complement component I, complement control protein factor I, complement factor I, complement factor I heavy chain, EC 3.4.21, EC 3.4.21.45, FIC3b-INA, I factor (complement), IF, KAF, Konglutinogen-activating factor, light chain of factor I
Host Species Mouse
Immunogen IF (NP_000195, 19 a.a. ~ 118 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KVTYTSQEDLVEKKCLAKKYTHLSCDKVFCQPWQRCIEGTCVCKLPYQCPKNGTAVCATNRRSFPTYCQQKSLECLHPGTKFLNNGTCTAEGKFSVSLKH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3426
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.