missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor H-related 4/CFHR4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18341401 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mL
1 ml
Unit Size:
0.25mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341401 0.25 mL 0.25mL
18607931 1 ml 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18341401 Supplier Novus Biologicals Supplier No. NBP2773780.25ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Complement Factor H-related 4/CFHR4 Polyclonal specifically detects Complement Factor H-related 4/CFHR4 in Human samples. It is validated for Western Blot, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor H-related 4/CFHR4
Applications Western Blot, Immunoassay
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered solution in PBS and 0.1% BSA
Gene Alias complement factor H-related protein 4, CFHL4, complement factor H-related 4, FHR4, FHR-4
Host Species Rabbit
Immunogen Recombinant FHR4 fragment SCR2-4.
Purification Method IgG purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10877
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.