missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H-related 2/CFHR2/CFHL2 Rabbit anti-Human, Clone: 5, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £429.00
Specifications
| Antigen | Complement Factor H-related 2/CFHR2/CFHL2 |
|---|---|
| Clone | 5 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222407
|
Novus Biologicals
NBP2-90262-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18267417
|
Novus Biologicals
NBP2-90262-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Factor H-related 2/CFHR2/CFHL2 Monoclonal specifically detects Complement Factor H-related 2/CFHR2/CFHL2 in Human samples. It is validated for ELISA.Specifications
| Complement Factor H-related 2/CFHR2/CFHL2 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 3080 | |
| IgG | |
| Protein A purified |
| 5 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CFHL2FHR-2, complement factor H-related 2, factor H-related gene 2, FHR2DDESK59, H factor (complement)-like 3, H factor-like 3, H factor-like protein 2, HFL3complement factor H-related protein 2 | |
| Recombinant Human Complement Factor H-related 2/CFHR2/CFHL2 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title