missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H Mouse anti-Human, Clone: C18/3, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23542-0.025MG
This item is not returnable.
View return policy
Description
Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Complement Factor H | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3075 | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| C18/3 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| CFH | |
| Human Factor H | |
| 0.025 mg | |
| Primary | |
| Clone C18/3 is specific for complement factor H in human serum and plasma | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction