missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H Antibody (L20/3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05125IR
This item is not returnable.
View return policy
Description
Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Complement Factor H | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CFH | |
| complement factor H in human serum and plasma (1) | |
| 0.1 mL | |
| Primary | |
| Specific for complement factor H in human serum and plasma (1). | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| L20/3 | |
| Western Blot, ELISA | |
| adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3075 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction