Learn More
Complement Factor H Antibody (L20/3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-05125-0.025MG
 View more versions of this product
Product Code. 18281243
  • £175.00 / 25µg
Description
Complement Factor H Monoclonal specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complement Factor H
Monoclonal
1.0 mg/mL
Western Blot 1:100-1:2000, ELISA 1:16000
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Mouse
Protein A or G purified
RUO
3075
Human
Purified
ELISA, Western Blot
L20/3
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
CFH
complement factor H in human serum and plasma (1)
0.025 mg
Primary
Specific for complement factor H in human serum and plasma (1).
Store at 4C. Do not freeze.
IgG1 κ