missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor H Antibody (C18/3), Biotin, Novus Biologicals™

Product Code. 18424852 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424852 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424852 Supplier Novus Biologicals Supplier No. NBP223542B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Factor H Monoclonal antibody specifically detects Complement Factor H in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor H
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone C18/3
Conjugate Biotin
Formulation PBS
Gene Alias adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Host Species Mouse
Immunogen Human Factor H (1)
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3075
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.