missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement factor D-IN-1, MedChemExpress
Description
Complement factor D-IN-1 is a potent and selective small-molecule reversible factor d inhibitor, with IC50s of 0.006 and 0.05 μM in FD Thioesterolytic Fluorescent Assay and a MAC Deposition Assay, respectively.
Specifications
Specifications
| Chemical Name or Material | Complement factor D-IN-1 |
| CAS | 1386455-76-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H19BrN6O3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 250 mg/mL (517.26 mM; Need ultrasonic) |
| SMILES | O=C(N)C1=NN(CC(N2[C@H](C(NC3=CC=CC(Br)=N3)=O)C[C@@H]4[C@H]2C4)=O)C5=C1C=CC=C5 |
| Molecular Weight (g/mol) | 483.32 |
| Formula Weight | 483.32 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?