missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor D/Adipsin Antibody (I8/1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05121AF700
This item is not returnable.
View return policy
Description
Complement Factor D/Adipsin Monoclonal specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Complement Factor D/Adipsin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CFD | |
| Human factor D. | |
| 0.1 mL | |
| Immunology | |
| 1675 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| I8/1 | |
| Western Blot, ELISA | |
| Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Clone I8/1 is specific for complement factor D in human serum and plasma | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction