missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor D/Adipsin Antibody (110), DyLight 550, Novus Biologicals™

Product Code. 18381791 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381791 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381791 Supplier Novus Biologicals Supplier No. NBP290630R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Complement Factor D/Adipsin Monoclonal specifically detects Complement Factor D/Adipsin in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor D/Adipsin
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Complement Factor D/Adipsin (P03953-2; Met1-Ser258).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1675
Test Specificity Has cross-reactivity in ELISA with: Human CFAD No cross-reactivity in ELISA with: Human CFH
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.