missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor D/Adipsin Antibody (005), DyLight 680, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Complement Factor D/Adipsin Monoclonal antibody specifically detects Complement Factor D/Adipsin in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Complement Factor D/Adipsin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 005 |
| Conjugate | DyLight 680 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Factor D/Adipsin (P00746; Ile26-Ala253). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?