missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement Factor B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551640
This item is not returnable.
View return policy
Description
Immunogen sequence: KDNEQHVFKV KDMENLEDVF YQMIDESQSL SLCGMVWEHR KGTDYHKQPW QAKISVIRPS KGHESCMGAV VSEYFVLTAA HCFTVDDKEH SIKVSVGGEK RDLEIEVVLF HPNYNINGKK EAGIPEFYDY DVALIKLKNK LKYG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 81%.
Factor B is a component of the alternative complement pathway. The protein is a single chain polypeptide with a molecular mass of 93 kDa. Factor B is cleaved by factor D into the Ba component (30 kDa) and the Bb (63 kDa). Bb together with C3b forms the alternative pathway C3 convertase C3bBb.
Specifications
| Complement Factor B | |
| Polyclonal | |
| Unconjugated | |
| CFB | |
| AHUS4; AI195813; AI255840; alternative-complement pathway C3/C5 convertase; ARMD14; B; BF; B-factor, properdin; BFD; C2; C3 proaccelerator; C3 proactivator; C3/C5 convertase; CFAB; CFB; CFBD; complement component 2 (within H-2S); complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; Da1-24; DADB-122G4.5; Factor B; Fb; FBI12; GBG; glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; H2-Bf; histocompatibility 2, complement component factor B; PBF2; properdin; properdin factor B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 629 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00751 | |
| CFB | |
| Recombinant protein corresponding to Human Complement Factor B. Recombinant protein control fragment (Product #RP-100390). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction