missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor B Antibody (SR1011), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22367-100ul
This item is not returnable.
View return policy
Description
Complement Factor B Monoclonal antibody specifically detects Complement Factor B in Human samples. It is validated for Western Blot
Specifications
| Complement Factor B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1011 | |
| Western Blot 1:500-1:2000 | |
| BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B | |
| A synthesized peptide derived from human Complement Factor B (Uniprot #: P00751) | |
| 100 μg | |
| Immunology | |
| 629 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction